1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)piperazine ≥95%

Supplier: Ambeed
Warning

Synonyms: 4-(Piperazino)methylphenylboronic acid pinacol ester

A858558-1G A858558-5G A858558-10G A858558-250MG
77614-534EA 104.03 USD
77614-534 77614-536 77686-899 77686-900
1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)piperazine ≥95%
1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)piperazine

Precaución: for Research Use Only

Formula: C₁₇H₂₇BN₂O₂
MW: 302.219 g/mol
MDL Number: MFCD18775799
CAS Number: 1245505-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR