N,N-Dimethyl-4-(2-(2-(octyloxy)phenyl)-6-phenylpyridin-4-yl)aniline 95+%

Supplier: Ambeed
Warning

A858197-5G A858197-100G A858197-25G
77614-532EA 16.48 USD
77614-532 77886-845 77886-846
N,N-Dimethyl-4-(2-(2-(octyloxy)phenyl)-6-phenylpyridin-4-yl)aniline 95+%
N,N-Dimethyl-4-(2-(2-(octyloxy)phenyl)-6-phenylpyridin-4-yl)aniline

Precaución: for Research Use Only

Formula: C₃₃H₃₈N₂O
MDL Number: MFCD11113565
CAS Number: 144190-25-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR