(S)-3-(Piperidin-2-yl)pyridine ≥97%

Supplier: Ambeed
Warning

Synonyms: Anabazin, (S)-Anabasine, (+)-Anabasine, (S)-3-(Piperidin-2-yl)pyridine

A857821-100MG A857821-250MG
77603-164EA 195.78 USD
77603-164 77627-668
(S)-3-(Piperidin-2-yl)pyridine ≥97%
(S)-3-(Piperidin-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂
MW: 162.23 g/mol
MDL Number: MFCD00871391
CAS Number: 494-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR