2-Oxa-7-azaspiro[3.5]nonane ≥98%

Supplier: Ambeed
Danger

A856245-1G A856245-100MG A856245-250MG A856245-5G
77573-722EA 441 USD
77573-722 77873-319 77873-320 77873-321
2-Oxa-7-azaspiro[3.5]nonane ≥98%
2-Oxa-7-azaspiro[3.5]nonane

Precaución: for Research Use Only

Formula: C₇H₁₃NO
MW: 127.18 g/mol
MDL Number: MFCD11035888
CAS Number: 241820-91-7
UN: 2735
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR