6-(Hydroxymethyl)uracil ≥98%

Supplier: Ambeed
Warning

A855822-250MG A855822-25G A855822-10G A855822-1G A855822-5G
77784-243EA 8 USD
77784-243 77784-244 77784-241 77784-242 77784-245
6-(Hydroxymethyl)uracil ≥98%
6-(Hydroxymethyl)uracil

Precaución: for Research Use Only

Formula: C₅H₆N₂O₃
MW: 142.11 g/mol
MDL Number: MFCD00797681
CAS Number: 22126-44-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR