Cerotic acid ≥98%

Supplier: Ambeed
Warning

A855633-1G A855633-250MG A855633-5G
77573-718EA 103 USD
77573-718 77618-406 77618-408
Cerotic acid ≥98%
Cerotic acid

Precaución: for Research Use Only

Formula: C₂₆H₅₂O₂
MDL Number: MFCD00002811
CAS Number: 506-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR