Lx1606 98+%

Supplier: Ambeed
Warning

Synonyms: Telotristat ethyl, Ethyl (S)-2-amino-3-(4-(2-amino-6-((R)-1-(4-chloro-2-(3-methyl-1H-pyrazol-1-yl)phenyl)-2,2,2-trifluoroethoxy)pyrimidin-4-yl)phenyl)propanoate

A855563-1MG A855563-5MG A855563-10MG A855563-100MG
77618-402EA 44.29 USD
77618-402 77618-404 77618-400 77686-896
Lx1606 98+%
Lx1606

Precaución: for Research Use Only

Formula: C₂₇H₂₆ClF₃N₆O₃
MW: 574.98 g/mol
MDL Number: MFCD18251583
CAS Number: 1033805-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR