(1R)-5,5',6,6',7,7',8,8'-Octahydro-[1,1'-Binaphthalene]-2,2'-diamine ≥97%

Supplier: Ambeed
Warning

A855494-1G A855494-250MG A855494-5G A855494-100MG
77618-394EA 146.26 USD
77618-394 77618-396 77618-398 77747-621
(1R)-5,5',6,6',7,7',8,8'-Octahydro-[1,1'-Binaphthalene]-2,2'-diamine ≥97%
(1R)-5,5',6,6',7,7',8,8'-Octahydro-[1,1'-Binaphthalene]-2,2'-diamine

Precaución: for Research Use Only

Formula: C₂₀H₂₄N₂
MW: 292.41 g/mol
CAS Number: 208248-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR