1,3,5-Tris(Aminomethyl)benzene trihydrochloride ≥95%

Supplier: Ambeed
Warning

Synonyms: Benzene-1,3,5-triyltrimethanamine trihydrochloride

A854937-250MG A854937-5G
77893-659EA 47 USD
77893-659 77893-660
1,3,5-Tris(Aminomethyl)benzene trihydrochloride ≥95%
1,3,5-Tris(Aminomethyl)benzene trihydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₈Cl₃N₃
MW: 274.62 g/mol
MDL Number: MFCD22628759
CAS Number: 69146-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR