2,3-Dibromo-5,6-dimethylpyridine ≥98%

Supplier: Ambeed
Warning

A854245-25G A854245-5G
77614-520EA 544.87 USD
77614-520 77684-815
2,3-Dibromo-5,6-dimethylpyridine ≥98%
2,3-Dibromo-5,6-dimethylpyridine

Precaución: For research use only.

Formula: C₇H₇Br₂N
MW: 264.945 g/mol
MDL Number: MFCD13688994
CAS Number: 117846-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR