Thieno[3,2-c]pyridin-4(5H)-one ≥95%

Supplier: Ambeed
Warning

A853955-5G A853955-1G A853955-250MG
77686-891EA 350 USD
77686-891 77901-937 77900-565
Thieno[3,2-c]pyridin-4(5H)-one ≥95%
Thieno[3,2-c]pyridin-4(5H)-one

Precaución: for Research Use Only

Formula: C₇H₅NOS
MDL Number: MFCD00673989
CAS Number: 27685-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR