1-(Triisopropylsilyl)-1-propyne ≥95%

Supplier: Ambeed
Warning

A853676-1G A853676-5G
77618-390EA 36.05 USD
77618-390 77749-661
1-(Triisopropylsilyl)-1-propyne ≥95%
1-(Triisopropylsilyl)-1-propyne

Precaución: for Research Use Only

Formula: C₁₂H₂₄Si
MDL Number: MFCD00015636
CAS Number: 82192-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR