2,4,6-Tribromophenyl methacrylate ≥97%

Supplier: Ambeed
Warning

A853046-1G A853046-5G
77595-368EA 25.63 USD
77595-368 77880-662
2,4,6-Tribromophenyl methacrylate ≥97%
2,4,6-Tribromophenyl methacrylate

Precaución: for Research Use Only

Formula: C₁₀H₇Br₃O₂
MDL Number: MFCD00078329
CAS Number: 37721-71-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR