1-Methyl-1H-indazole-5-boronic acid pinacol ester ≥98%

Supplier: Ambeed
Warning

A852445-1G A852445-25G
77884-784EA 50 USD
77884-784 77884-785
1-Methyl-1H-indazole-5-boronic acid pinacol ester ≥98%
1-Methyl-1H-indazole-5-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₉BN₂O₂
MDL Number: MFCD15071438
CAS Number: 1235469-00-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR