2-Bromo-3-methylisonicotinonitrile ≥97%

Supplier: Ambeed
Warning

A852042-1G A852042-250MG
77618-386EA 251.47 USD
77618-386 77747-619
2-Bromo-3-methylisonicotinonitrile ≥97%
2-Bromo-3-methylisonicotinonitrile

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
MDL Number: MFCD28665862
CAS Number: 1260024-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR