3-Oxo-1-oxa-7-azaspiro[4.4]nonane-7-carboxylic acid tert-butyl ester ≥97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-oxo-1-oxa-7-azaspiro[4.4]nonane-7-carboxylate

A851027-1G A851027-250MG
77674-256EA 349 USD
77674-256 77884-782
3-Oxo-1-oxa-7-azaspiro[4.4]nonane-7-carboxylic acid tert-butyl ester ≥97%
3-Oxo-1-oxa-7-azaspiro[4.4]nonane-7-carboxylic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO₄
MW: 241.29 g/mol
MDL Number: MFCD12198523
CAS Number: 1160246-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR