(2S,4S)-4-Fluoro-1-((2-methyl-4-oxo-4-(pyrrolidin-1-yl)butan-2-yl)glycyl)pyrrolidine-2-carbonitrile 98+%

Supplier: Ambeed
Warning

Synonyms: DBPR108

A850169-10MG A850169-1MG A850169-5MG
77618-376EA 48.41 USD
77618-376 77618-378 77618-380
(2S,4S)-4-Fluoro-1-((2-methyl-4-oxo-4-(pyrrolidin-1-yl)butan-2-yl)glycyl)pyrrolidine-2-carbonitrile 98+%
(2S,4S)-4-Fluoro-1-((2-methyl-4-oxo-4-(pyrrolidin-1-yl)butan-2-yl)glycyl)pyrrolidine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₁₆H₂₅FN₄O₂
MW: 324.39 g/mol
MDL Number: MFCD28385883
CAS Number: 1186426-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR