1,6-Hexylenediphosphonic acid ≥99%

Supplier: Ambeed
Danger

A849622-1G A849622-250MG
77618-372EA 221.45 USD
77618-372 77747-617
1,6-Hexylenediphosphonic acid ≥99%
1,6-Hexylenediphosphonic acid

Precaución: for Research Use Only

Formula: C₆H₁₆O₆P₂
MDL Number: MFCD01630742
CAS Number: 4721-22-6
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR