4-tert-Butyltoluene ≥99%

Supplier: Ambeed
Danger

A849329-100G A849329-25G
77599-174EA 20.5 USD
77599-174 77878-696
4-tert-Butyltoluene ≥99%
4-tert-Butyltoluene

Precaución: for Research Use Only

Formula: C₁₁H₁₆
MDL Number: MFCD00008837
CAS Number: 98-51-1
UN: 1993
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR