5-Chlorothiophene-2-carbonitrile ≥95%

Supplier: Ambeed
Danger

A848840-1G A848840-5G
77670-218EA 26 USD
77670-218 77873-311
5-Chlorothiophene-2-carbonitrile ≥95%
5-Chlorothiophene-2-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₂ClNS
MDL Number: MFCD09265032
CAS Number: 50478-16-5
UN: 2922
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR