2',4'-Dimethoxy-2,2,2-trifluoroacetophenone ≥97%

Supplier: Ambeed
Warning

A848395-100MG A848395-1G A848395-250MG
77880-651EA 25 USD
77880-651 77880-652 77880-653
2',4'-Dimethoxy-2,2,2-trifluoroacetophenone ≥97%
2',4'-Dimethoxy-2,2,2-trifluoroacetophenone

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₃
MDL Number: MFCD00835330
CAS Number: 578-16-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR