1-(Trifluoromethyl)cyclobutan-1-amine hydrochloride ≥95%

Supplier: Ambeed
Warning

A848317-100MG A848317-250MG
77595-360EA 103.53 USD
77595-360 77614-518
1-(Trifluoromethyl)cyclobutan-1-amine hydrochloride ≥95%
1-(Trifluoromethyl)cyclobutan-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₉ClF₃N
MW: 175.58 g/mol
MDL Number: MFCD15526688
CAS Number: 1260768-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR