1,4-Diaminonaphthalene ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: Naphthalene-1,4-diamine

A847863-100MG A847863-250MG
77684-811EA 20 USD
77684-811 77891-602
1,4-Diaminonaphthalene ≥98%, Ambeed.Inc
1,4-Diaminonaphthalene

Precaución: For research use only.

Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
MDL Number: MFCD01712813
CAS Number: 2243-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR