Betaine ≥96%

Supplier: Ambeed
Warning

A847788-1000G A847788-100G A847788-25G A847788-500G
77595-354EA 56.38 USD
77595-354 77595-356 77618-368 77618-370
Betaine ≥96%
Betaine

Precaución: for Research Use Only

Formula: C₅H₁₁NO₂
MDL Number: MFCD00012123
CAS Number: 107-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR