6-Chloro-3-oxo-2,3-dihydropyridazine-4-carboxylic acid methyl ester ≥98%

Supplier: Ambeed
Warning

A847588-100MG A847588-1G A847588-250MG A847588-5G
77886-835EA 27 USD
77886-835 77886-836 77886-837 77886-838
6-Chloro-3-oxo-2,3-dihydropyridazine-4-carboxylic acid methyl ester ≥98%
6-Chloro-3-oxo-2,3-dihydropyridazine-4-carboxylic acid methyl ester

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₃
MDL Number: MFCD16250585
CAS Number: 89581-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR