Potassium 2-fluorobenzyl-trifluoroborate ≥97%

Supplier: Ambeed
Warning

Synonyms: Potassium trifluoro-[(2-fluorophenyl)methyl]boranuide, Potassium trifluoro[(2-fluorophenyl)methyl]boranuide

A847067-5G A847067-100MG A847067-1G
77784-223EA 654 USD
77784-223 77784-221 77784-222
Potassium 2-fluorobenzyl-trifluoroborate ≥97%
Potassium 2-fluorobenzyl-trifluoroborate

Precaución: for Research Use Only

Formula: C₇H₆BF₄·K
MW: 216.025 g/mol
MDL Number: MFCD09993122
CAS Number: 1246073-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR