Phenyl-2,3,4,6-tetra-O-acetyl-1-thio-β-D-galactopyranoside ≥98%

Supplier: Ambeed
Warning

A846938-1G A846938-25G A846938-5G
77670-212EA 50 USD
77670-212 77880-648 77880-649
Phenyl-2,3,4,6-tetra-O-acetyl-1-thio-β-D-galactopyranoside ≥98%
Phenyl-2,3,4,6-tetra-O-acetyl-1-thio-beta-D-galactopyranoside

Precaución: for Research Use Only

Formula: C₂₀H₂₄O₉S
MDL Number: MFCD06657849
CAS Number: 24404-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR