Di-tert-butyl succinate ≥95%

Supplier: Ambeed
Warning

A846191-1G A846191-250MG A846191-5G
77686-876EA 82 USD
77686-876 77686-877 77686-878
Di-tert-butyl succinate ≥95%
Di-tert-butyl succinate

Precaución: for Research Use Only

Formula: C₁₂H₂₂O₄
MW: 230.30 g/mol
MDL Number: MFCD00127979
CAS Number: 926-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR