Potassium (4-chlorobenzyl)trifluoroborate ≥98%

Supplier: Ambeed
Warning

A845017-100MG A845017-1G A845017-250MG
77623-036EA 140.08 USD
77623-036 77784-210 77784-211
Potassium (4-chlorobenzyl)trifluoroborate ≥98%
Potassium (4-chlorobenzyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₇H₆BClF₃K
MW: 232.48 g/mol
MDL Number: MFCD09993369
CAS Number: 1334209-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR