Methyl 5,6-dichloropyridazine-3-carboxylate ≥95%

Supplier: Ambeed
Warning

A844965-5G A844965-1G A844965-100MG
77749-649EA 1977 USD
77749-649 77749-648 77749-647
Methyl 5,6-dichloropyridazine-3-carboxylate ≥95%
Methyl 5,6-dichloropyridazine-3-carboxylate

Precaución: for Research Use Only

Formula: C₆H₄Cl₂N₂O₂
MW: 207.01 g/mol
MDL Number: MFCD31412838
CAS Number: 1591827-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR