tert-Butyl methyl(3-(methylamino)propyl)carbamate ≥95%

Supplier: Ambeed
Danger

A844674-5G A844674-100MG A844674-1G
77599-168EA 499.18 USD
77599-168 77784-208 77784-209
tert-Butyl methyl(3-(methylamino)propyl)carbamate ≥95%
tert-Butyl methyl(3-(methylamino)propyl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₂₂N₂O₂
MDL Number: MFCD14155832
CAS Number: 123183-72-2
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR