4-((2,4-Diaminopyrimidin-5-yl)methyl)-2,6-dimethoxyphenol ≥97%

Supplier: Ambeed
Warning

A844334-1G A844334-100MG
77573-682EA 280 USD
77573-682 77618-360
4-((2,4-Diaminopyrimidin-5-yl)methyl)-2,6-dimethoxyphenol ≥97%
4-((2,4-Diaminopyrimidin-5-yl)methyl)-2,6-dimethoxyphenol

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₄O₃
MDL Number: MFCD31536269
CAS Number: 21253-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR