[(S,S)-Teth-TsDPEN-RuCl] ≥98%

Supplier: Ambeed
Warning

Synonyms: {N-[3-(η6-phenyl)propyl]-[(1S-2S)-1,2-diphenyl-1-4-methylbenzenesulphonylamidato(kN)-ethyl-2-amino-(kN)]}ruthenium(II)

A843814-100MG A843814-1G A843814-5G A843814-250MG
77618-356EA 66.95 USD
77618-356 77618-358 77747-609 77747-608
[(S,S)-Teth-TsDPEN-RuCl] ≥98%
[(S,S)-Teth-TsDPEN-RuCl]

Precaución: for Research Use Only

Formula: C₃₀H₃₁ClN₂O₂RuS
MW: 620.17 g/mol
MDL Number: MFCD16294984
CAS Number: 851051-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR