(S)-2-(2-Amino-5-chlorophenyl)-4-cyclopropyl-1,1,1-trifluorobut-3-yn-2-ol ≥95%

Supplier: Ambeed
Warning

A842986-1G A842986-5G
77882-765EA 49 USD
77882-765 77882-766
(S)-2-(2-Amino-5-chlorophenyl)-4-cyclopropyl-1,1,1-trifluorobut-3-yn-2-ol ≥95%
(S)-2-(2-Amino-5-chlorophenyl)-4-cyclopropyl-1,1,1-trifluorobut-3-yn-2-ol

Precaución: for Research Use Only

Formula: C₁₃H₁₁ClF₃NO
MDL Number: MFCD09751012
CAS Number: 209414-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR