tert-Butyl (R)-2-methyl-3-oxoazetidine-1-carboxylate ≥97%

Supplier: Ambeed
Danger

A841769-250MG A841769-1G
77749-646EA 241 USD
77749-646 77749-645
tert-Butyl (R)-2-methyl-3-oxoazetidine-1-carboxylate ≥97%
tert-Butyl (R)-2-methyl-3-oxoazetidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₅NO₃
MW: 185.22 g/mol
MDL Number: MFCD28501894
CAS Number: 1638744-93-0
UN: 3335
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR