(R)-(-)-2-Fluoro-α-methyl-4-biphenylacetic acid ≥98%

Supplier: Ambeed
Danger

Synonyms: Tarenflurbil, (R)-2-(2-Fluoro-[1,1'-biphenyl]-4-yl)propanoic acid, (R)-(-)-Flurbiprofen

A841572-50MG A841572-100MG A841572-250MG
77614-516EA 24.72 USD
77614-516 77674-246 77674-248
(R)-(-)-2-Fluoro-α-methyl-4-biphenylacetic acid ≥98%
(R)-(-)-2-Fluoro-α-methyl-4-biphenylacetic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₂
MW: 244.260 g/mol
MDL Number: MFCD00869714
CAS Number: 51543-40-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR