5-Iodo-2-methylanisole ≥98%

Supplier: Ambeed
Warning

A840939-1G A840939-250MG
77595-342EA 301.35 USD
77595-342 77784-194
5-Iodo-2-methylanisole ≥98%
5-Iodo-2-methylanisole

Precaución: for Research Use Only

Formula: C₈H₉IO
MDL Number: MFCD06797971
CAS Number: 220728-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR