(3,5-Dichloropyridin-2-yl)boronic acid 95+%

Supplier: Ambeed
Warning

A840600-1G A840600-250MG
77886-832EA 317 USD
77886-832 77886-833
(3,5-Dichloropyridin-2-yl)boronic acid 95+%
(3,5-Dichloropyridin-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₅H₄BCl₂NO₂
MDL Number: MFCD09028487
CAS Number: 1309982-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR