Plinabulin ≥98%

Supplier: Ambeed
Warning

Synonyms: (3Z,6Z)-3-benzylidene-6-[(5-tert-butyl-1H-imidazol-4-yl)methylidene]piperazine-2,5-dione, NPI-2358, (Z)-3-((Z)-Benzylidene)-6-((5-(tert-butyl)-1H-imidazol-4-yl)methylene)piperazine-2,5-dione

A840538-10MG A840538-1MG A840538-5MG
77614-510EA 76.22 USD
77614-510 77614-512 77614-514
Plinabulin ≥98%
Plinabulin

Precaución: for Research Use Only

Formula: C₁₉H₂₀N₄O₂
MW: 336.39 g/mol
MDL Number: MFCD18074510
CAS Number: 714272-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR