2,5-Dioxopyrrolidin-1-yl 1-(9H-fluoren-9-yl)-3-oxo-2,7,10,13,16-pentaoxa-4-azanonadecan-19-oate ≥95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-PEG4-NHS ester
A839313-250MG
A839313-100MG
77614-506EA
37.08
USD
77614-506
77684-800
2,5-Dioxopyrrolidin-1-yl 1-(9H-fluoren-9-yl)-3-oxo-2,7,10,13,16-pentaoxa-4-azanonadecan-19-oate ≥95%
2,5-Dioxopyrrolidin-1-yl 1-(9H-fluoren-9-yl)-3-oxo-2,7,10,13,16-pentaoxa-4-azanonadecan-19-oate
Precaución: For research use only.
Formula:
C₃₀H₃₆N₂O₁₀ MW: 584.61 g/mol |
MDL Number:
MFCD20227659 CAS Number: 1314378-14-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...