2,3-Dihydro-1H-benzo[e]indole ≥95%

Supplier: Ambeed
Warning

A839188-100MG A839188-250MG
77749-640EA 187 USD
77749-640 77749-641
2,3-Dihydro-1H-benzo[e]indole ≥95%
2,3-Dihydro-1H-benzo[e]indole

Precaución: for Research Use Only

Formula: C₁₂H₁₁N
MW: 169.22 g/mol
MDL Number: MFCD20664337
CAS Number: 5811-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR