(2S)-2-Amino-2-cyclopropylethan-1-ol hydrochloride ≥95%

Supplier: Ambeed
Warning

Synonyms: (2S)-2-amino-2-cyclopropylethan-1-ol hcl

A838693-250MG A838693-1G
77599-156EA 95.33 USD
77599-156 77623-016
(2S)-2-Amino-2-cyclopropylethan-1-ol hydrochloride ≥95%
(2S)-2-Amino-2-cyclopropylethan-1-ol hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₁NO·ClH
MW: 137.608 g/mol
MDL Number: MFCD28975203
CAS Number: 1623432-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR