1-(tert-Butyl)-3,5-dichlorobenzene ≥95%

Supplier: Ambeed
Danger

A838375-1G A838375-5G A838375-25G
77573-664EA 20 USD
77573-664 77595-334 77784-187
1-(tert-Butyl)-3,5-dichlorobenzene ≥95%
1-(tert-Butyl)-3,5-dichlorobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂Cl₂
MDL Number: MFCD28466945
CAS Number: 1369899-62-6
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR