20-Methoxy-20-oxoicosanoic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: 20-Methoxy-20-oxoicosanoicacid

A838296-1G A838296-250MG
77623-008EA 47.38 USD
77623-008 77623-010
20-Methoxy-20-oxoicosanoic acid ≥98%
20-Methoxy-20-oxoicosanoic acid

Precaución: for Research Use Only

Formula: C₂₁H₄₀O₄
MW: 356.5399 g/mol
MDL Number: MFCD32196848
CAS Number: 1767-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR