N-(8-Amino-6-fluoro-5-methyl-1-oxo-1,2,3,4-tetrahydronaphthalen-2-yl)acetamide ≥98%

Supplier: Ambeed
Warning

Synonyms: Exatecan Intermediate 2

A838144-250MG A838144-10G
77618-340EA 60.77 USD
77618-340 77749-638
N-(8-Amino-6-fluoro-5-methyl-1-oxo-1,2,3,4-tetrahydronaphthalen-2-yl)acetamide ≥98%
N-(8-Amino-6-fluoro-5-methyl-1-oxo-1,2,3,4-tetrahydronaphthalen-2-yl)acetamide

Precaución: for Research Use Only

Formula: C₁₃H₁₅FN₂O₂
MW: 250.27 g/mol
MDL Number: MFCD18827384
CAS Number: 182182-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR