2-((tert-Butoxycarbonyl)(cyclopropyl)amino)acetic acid ≥95%

Supplier: Ambeed
Danger

Synonyms: N-(tert-Butoxycarbonyl)-N-cyclopropylglycine

A836634-250MG A836634-5G
77686-863EA 48 USD
77686-863 77686-864
2-((tert-Butoxycarbonyl)(cyclopropyl)amino)acetic acid ≥95%
2-((tert-Butoxycarbonyl)(cyclopropyl)amino)acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₄
MW: 215.25 g/mol
MDL Number: MFCD12158392
CAS Number: 182291-93-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR