6-Oxaspiro[2.5]octane-1-carboxylic acid ≥98%

Supplier: Ambeed
Warning

A836586-100MG A836586-1G
77893-629EA 69 USD
77893-629 77893-630
6-Oxaspiro[2.5]octane-1-carboxylic acid ≥98%
6-Oxaspiro[2.5]octane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₂O₃
MDL Number: MFCD16744613
CAS Number: 909406-73-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR