(S)-2,2'-Bis(bromomethyl)-1,1'-binaphthalene ≥98%

Supplier: Ambeed
Danger

A836299-1G A836299-250MG
77684-797EA 218 USD
77684-797 77684-798
(S)-2,2'-Bis(bromomethyl)-1,1'-binaphthalene ≥98%
(S)-2,2'-Bis(bromomethyl)-1,1'-binaphthalene

Precaución: For research use only.

Formula: C₂₂H₁₆Br₂
MW: 440.17 g/mol
MDL Number: MFCD00185730
CAS Number: 37803-02-4
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR