4-Methyl-1,3-dihydro-2H-benzimidazol-2-one ≥97%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-1H-benzo[d]imidazol-2(3H)-one

A835953-1G A835953-250MG
77884-774EA 10 USD
77884-774 77884-775
4-Methyl-1,3-dihydro-2H-benzimidazol-2-one ≥97%
4-Methyl-1,3-dihydro-2H-benzimidazol-2-one

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MDL Number: MFCD08274669
CAS Number: 19190-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR