Sodium-D(+)-glucuronate monohydrate ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
D-Glucuronic acid sodium salt hydrate, D-Glucuronic acid sodium salt monohydrate, D(+)-Glucuronic acid, sodium salt, monohydrate, Sodium (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoate hydrate
A835541-25G
A835541-5G
A835541-100G
A835541-1G
77603-118EA
44.08
USD
77603-118
77603-120
77627-640
77880-637
Sodium-D(+)-glucuronate monohydrate ≥98%
Sodium D(+)-glucuronate monohydrate
Precaución: for Research Use Only
Formula:
C₆H₁₀O₇·1H₂O MW: 234.14 g/mol (216.14 anhy) |
MDL Number:
MFCD00151051 CAS Number: 207300-70-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...